Learn More
Zymo Research Corporation Zymo-Spin™ IIICG Columns (50 Pack) (Capped) Green Retension Ring
SDP
Catalog No. NC0921397
Encompass_Preferred
The Zymo-Spin IIICG features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 100 g of RNA, in 50l eluate. Capacity is 800 l.
Catalog No. NC0921397 Supplier Zymo Research Corporation Supplier No. C100650G
The Zymo-Spin IIICG features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 25 g of DNA, and 100 g of RNA, in 50l eluate. Capacity is 800 l.