Learn More
Zymo Research Corporation ZymoBIOMICS 96 MagBead DNA/RNA Kit (96 Preps.)
SDP
Catalog No. 501957610
Encompass_Preferred
The provided DNA/RNA Shield™ inactivates infectious agents and is ideal for sample storage at ambient temperatures. The extraction method utilizes magnetic beads for DNA/RNA extraction without the use of phenol and is eluted into 50 l of ZymoBIOMICS™ DNase/RNaseFree Water. DNA/RNA is ready for any downstream application including Next-Gen Sequencing, RT-qPCR, etc.
Catalog No. 50-195-7610 Supplier Zymo Research Corporation Supplier No. R2135
The provided DNA/RNA Shield™ inactivates infectious agents and is ideal for sample storage at ambient temperatures. The extraction method utilizes magnetic beads for DNA/RNA extraction without the use of phenol and is eluted into 50 l of ZymoBIOMICS™ DNase/RNaseFree Water. DNA/RNA is ready for any downstream application including Next-Gen Sequencing, RT-qPCR, etc.