Learn More
Zymo Research Corporation ZymoBIOMICS 96 MagBead DNA/RNA Kit (4 x 96 Preps.)
SDP
Catalog No. 501957611
Encompass_Preferred
The ZymoBIOMICS MagBead DNA/RNA kit provides a high-throughput, magnetic bead-based purification of both high-quality DNA and total RNA (including small/microRNAs) from the same starting sample. The provided DNA/RNA Shield inactivates infectious agents and is ideal for sample storage at ambient temperatures. The extraction method utilizes magnetic beads for DNA/RNA extraction without the use of phenol and is eluted into ≥50 µl of ZymoBIOMICS DNase/RNase-Free Water. DNA/RNA is ready for any downstream application including Next-Gen Sequencing, RT-qPCR, etc.
Catalog No. 50-195-7611 Supplier Zymo Research Corporation Supplier No. R2136
The ZymoBIOMICS MagBead DNA/RNA kit provides a high-throughput, magnetic bead-based purification of both high-quality DNA and total RNA (including small/microRNAs) from the same starting sample. The provided DNA/RNA Shield inactivates infectious agents and is ideal for sample storage at ambient temperatures. The extraction method utilizes magnetic beads for DNA/RNA extraction without the use of phenol and is eluted into ≥50 µl of ZymoBIOMICS DNase/RNase-Free Water. DNA/RNA is ready for any downstream application including Next-Gen Sequencing, RT-qPCR, etc.