Learn More
Zymo Research Corporation ZymoBIOMICS DNA Microprep Kit (lysis Matrix Not Included) (50 preps)
SDP
Catalog No. 501251503
Encompass_Preferred
The ZymoBIOMICS DNA Microprep Kitis designed for purifying DNA from a wide array of sample inputs (e.g.feces, soil, water, and biofilms), that is immediately ready for microbiome or metagenome analyses.
Catalog No. 50-125-1503 Supplier Zymo Research Corporation Supplier No. D4305
The ZymoBIOMICS DNA Microprep Kitis designed for purifying DNA from a wide array of sample inputs (e.g.feces, soil, water, and biofilms), that is immediately ready for microbiome or metagenome analyses.