Learn More
Zymo Research Corporation ZymoBIOMICS DNA 96 MagBead Kit (2 x 96 preps)Includes: [D4306] ZymoBIOMICS 96 MagBead Kit x 1, [S6002-96-4] ZR BashingBead Lysis Racks (0.1mm & 0.5 mm) 2 pack x 1[D4300-1-150] ZymoBIOMICS Lysis Solution, 150 ml x 1
SDP
Catalog No. 501251494
Encompass_Preferred
The ZymoBIOMICS DNA Kits are designed for purifying DNA from a variety of sample inputs that is immediately ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. Gram-negative/positive bacteria, fungus, protozoans, and algae), making it ideal for microbial community profiling. Uniform mechanical lysis of all microbes is achieved by bead beating with the innovative ultra-high density BashingBeads. This kit is equipped with our OneStep PCR Inhibitor removal technology, enabling PCR amplification from DNA derived from inhibitor-rich environmental samples. Purified DNA is ideal for all downstream applications including PCR, arrays, 16S rRNA gene sequencing, and shotgun sequencing. DNA Size is 15-20 kb.
Catalog No. 50-125-1494 Supplier Zymo Research Corporation Supplier No. D4302
The ZymoBIOMICS DNA Kits are designed for purifying DNA from a variety of sample inputs that is immediately ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. Gram-negative/positive bacteria, fungus, protozoans, and algae), making it ideal for microbial community profiling. Uniform mechanical lysis of all microbes is achieved by bead beating with the innovative ultra-high density BashingBeads. This kit is equipped with our OneStep PCR Inhibitor removal technology, enabling PCR amplification from DNA derived from inhibitor-rich environmental samples. Purified DNA is ideal for all downstream applications including PCR, arrays, 16S rRNA gene sequencing, and shotgun sequencing. DNA Size is 15-20 kb.