Learn More
Zymo Research Corporation ZymoBIOMICS™ Lysis Solution, 150 ml
SDP
Catalog No. NC1420714
Encompass_Preferred
Used in combination with the ZR BashingBead Lysis Tubes, it assists with microbial lysis. It is certified to be low-bioburden to ensure minimal background contaminants for low abundance microbiome samples.
Catalog No. NC1420714 Supplier Zymo Research Corporation Supplier No. D43001150
Used in combination with the ZR BashingBead Lysis Tubes, it assists with microbial lysis. It is certified to be low-bioburden to ensure minimal background contaminants for low abundance microbiome samples.