Learn More
Zymo Research Corporation ZymoBIOMICS Lysis Solution, 40 ml
SDP
Catalog No. 501251489
Encompass_Preferred
Used in combination with the ZR BashingBead Lysis Tubes, it assists with microbial lysis. It is certified to be low-bioburden to ensure minimal background contaminants for low abundance microbiome samples.
Catalog No. 50-125-1489 Supplier Zymo Research Corporation Supplier No. D4300140
Used in combination with the ZR BashingBead Lysis Tubes, it assists with microbial lysis. It is certified to be low-bioburden to ensure minimal background contaminants for low abundance microbiome samples.