Learn More
Zymo Research Corporation ZymoBIOMICS™ MagBead (96 preps)
SDP
Catalog No. NC1835516
Encompass_Preferred
The provided DNA/RNA Shield™ inactivates infectious agents and is ideal for sample storage at ambient temperatures. The extraction method utilizes magnetic beads for DNA/RNA extraction without the use of phenol and is eluted into 50 l of ZymoBIOMICS™ DNase/RNase-Free Water. RNA is ready for any downstream application including Next-Gen Sequencing, RT-qPCR, etc.
Catalog No. NC1835516 Supplier Zymo Research Corporation Supplier No. R2137
The provided DNA/RNA Shield™ inactivates infectious agents and is ideal for sample storage at ambient temperatures. The extraction method utilizes magnetic beads for DNA/RNA extraction without the use of phenol and is eluted into 50 l of ZymoBIOMICS™ DNase/RNase-Free Water. RNA is ready for any downstream application including Next-Gen Sequencing, RT-qPCR, etc.