Learn More
Zymo Research Corporation ZymoBIOMICS™ RNA Mini Kit (50 preps)
SDP
Catalog No. 501251712
Encompass_Preferred
The ZymoBIOMICS RNA Miniprep Kit is designed for purifying RNA from a wide array of sample inputs that is ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. Gram-negative/positive bacteria, fungus, protozoans, and algae). The procedure uses Zymo-Spin Column technology that results in high-quality total RNA (including small RNAs 17-200 nt) that is free of PCR inhibitors and is ready for RT-PCR, hybridization, sequencing, etc. DNase I included.
Catalog No. 50-125-1712 Supplier Zymo Research Corporation Supplier No. R2001
The ZymoBIOMICS RNA Miniprep Kit is designed for purifying RNA from a wide array of sample inputs that is ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. Gram-negative/positive bacteria, fungus, protozoans, and algae). The procedure uses Zymo-Spin Column technology that results in high-quality total RNA (including small RNAs 17-200 nt) that is free of PCR inhibitors and is ready for RT-PCR, hybridization, sequencing, etc. DNase I included.