Learn More
Zymo Research Corporation ZymoBIOMICS Spike-in Control I (High Microbial Load) (500 µl x 10)
SDP
Catalog No. 501957592
Encompass_Preferred
When spiked into an unknown sample, this product will serve as an in situ positive control for DNAsequencing-based (e.g. NGS-based) microbiome measurements. These two bacteria, Imtechella halotolerans (Gram-negative) and Allobacillus halotolerans (Gram-positive) represent different cell recalcitrance and can expose potential bias during DNA extraction. Moreover, with accurately quantified cell numbers, this standard enables absolute cell number quantification in microbiome measurements.
Catalog No. 50-195-7592 Supplier Zymo Research Corporation Supplier No. D632010
When spiked into an unknown sample, this product will serve as an in situ positive control for DNAsequencing-based (e.g. NGS-based) microbiome measurements. These two bacteria, Imtechella halotolerans (Gram-negative) and Allobacillus halotolerans (Gram-positive) represent different cell recalcitrance and can expose potential bias during DNA extraction. Moreover, with accurately quantified cell numbers, this standard enables absolute cell number quantification in microbiome measurements.