Learn More
Research Products International Corp Zymoclean Gel DNA Recovery Kit w/ Zymo-Spin I Columns, Uncapped, 50 Preps
SDP
Catalog No. 501977910
Encompass
The Zymoclean Gel DNA Recovery Kit is a gel DNA extraction kit that provides rapid purification of high quality DNA from TAE/TBE-buffered agarose gels. This gel DNA extraction kit features Zymo-Spin technology to yield high-quality, purified DNA in just minutes. DNA purified using the Zymoclean Gel DNA Recovery kits is perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.
Quick recovery of ultra-pure DNA from agarose gels.
Column design permits DNA elution at high concentrations into minimal volumes.
Eluted DNA is well-suited for use in DNA ligation, sequencing, labeling, PCR, etc.
Catalog No. 50-197-7910 Supplier Research Products International Corp Supplier No. ZD4001
The Zymoclean Gel DNA Recovery Kit is a gel DNA extraction kit that provides rapid purification of high quality DNA from TAE/TBE-buffered agarose gels. This gel DNA extraction kit features Zymo-Spin technology to yield high-quality, purified DNA in just minutes. DNA purified using the Zymoclean Gel DNA Recovery kits is perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.
Quick recovery of ultra-pure DNA from agarose gels.
Column design permits DNA elution at high concentrations into minimal volumes.
Eluted DNA is well-suited for use in DNA ligation, sequencing, labeling, PCR, etc.