Learn More
Zymo Research Corporation Zymoclean™ Gel DNA Recovery Kit (50 Preps) w/ Zymo-Spin™ I Columns (Uncapped)
SDP
Catalog No. NC9681173
Encompass_Preferred
Simply add the specially formulated Agarose Dissolving Buffer (ADB) to the gel slice containing your DNA sample, let dissolve, and then transfer to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product utilizes Fast-Spin column technology to yield high-quality DNA in just 15 minutes. DNA purified using the Zymoclean™ Gel DNA Recovery Kit is perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.
Catalog No. NC9681173 Supplier Zymo Research Corporation Supplier No. D4001
Simply add the specially formulated Agarose Dissolving Buffer (ADB) to the gel slice containing your DNA sample, let dissolve, and then transfer to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product utilizes Fast-Spin column technology to yield high-quality DNA in just 15 minutes. DNA purified using the Zymoclean™ Gel DNA Recovery Kit is perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.