Learn More
Zymo Research Corporation Zymolyase (2000 Units Lyophilized) w/ Storage Buffer (500 µl)
SDP
Catalog No. 50444504
Encompass
The primary yeast lytic activity is -1,3 glucanase and -1,3-glucan laminaripentao-hydrolase. It hydrolyzes glucose polymers at the -1,3-glucan linkages releasing laminaripentaose as the principal product.
Catalog No. 50-444-504 Supplier Zymo Research Corporation Supplier No. E1005
The primary yeast lytic activity is -1,3 glucanase and -1,3-glucan laminaripentao-hydrolase. It hydrolyzes glucose polymers at the -1,3-glucan linkages releasing laminaripentaose as the principal product.