Learn More
Zymo Research Corporation Zymoprep™ -96 Yeast Plasmid Miniprep (2 x 96 preps)  [Includes E1004 x 2: Zymolyase (1 x 2000 Units Lyophilized) w/ Storage Buffer (500µl)]
SDP
Catalog No. 501251340
Encompass_Preferred
The Zymoprep™-96 Yeast Plasmid Miniprep kit provides all the necessary reagents for high-throughput plasmid isolation from S. cerevisiae, C. albicans, S. pombe and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The Zymoprep™-96 is a simple and efficient yeast plasmid miniprep kit that is based on the classic E. coli alkaline lysis method with our Zymolyase™ added in the first solution. There is no need for glass beads, or phenol.
Catalog No. 50-125-1340 Supplier Zymo Research Corporation Supplier No. D2005
The Zymoprep™-96 Yeast Plasmid Miniprep kit provides all the necessary reagents for high-throughput plasmid isolation from S. cerevisiae, C. albicans, S. pombe and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The Zymoprep™-96 is a simple and efficient yeast plasmid miniprep kit that is based on the classic E. coli alkaline lysis method with our Zymolyase™ added in the first solution. There is no need for glass beads, or phenol.