Learn More
Zymo Research Corporation Zymoprep™ -96 Yeast Plasmid Miniprep (8 x 96 preps)
SDP
Catalog No. 501251342
Encompass_Preferred
The Zymoprep™-96 Yeast Plasmid Miniprep kit provides all the necessary reagents for high-throughput plasmid isolation from S. cerevisiae, C. albicans, S. pombe and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The Zymoprep™-96 is a simple and efficient yeast plasmid miniprep kit that is based on the classic E. coli alkaline lysis method with our Zymolyase™ added in the first solution. There is no need for glass beads, or phenol.
Catalog No. 50-125-1342 Supplier Zymo Research Corporation Supplier No. D2007
The Zymoprep™-96 Yeast Plasmid Miniprep kit provides all the necessary reagents for high-throughput plasmid isolation from S. cerevisiae, C. albicans, S. pombe and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis. The Zymoprep™-96 is a simple and efficient yeast plasmid miniprep kit that is based on the classic E. coli alkaline lysis method with our Zymolyase™ added in the first solution. There is no need for glass beads, or phenol.