Learn More
Zymo Research Corporation ZymoPURE™ Binding Buffer (110 ml)
SDP
Catalog No. 501251465
Encompass_Preferred
ZymoPURE Binding Buffer is designed to be used with our ZymoPURE Plasmid Purification Kits, which is the fastest and simplest method available to efficiently isolate transfection-grade plasmid DNA from E. coli.
Catalog No. 50-125-1465 Supplier Zymo Research Corporation Supplier No. D42004110
ZymoPURE Binding Buffer is designed to be used with our ZymoPURE Plasmid Purification Kits, which is the fastest and simplest method available to efficiently isolate transfection-grade plasmid DNA from E. coli.