Learn More
Zymo Research Corporation ZymoPURE™ Binding Buffer (150 ml)
SDP
Catalog No. 501251466
Encompass_Preferred
ZymoPURE™ Binding Buffer (150 ml). Assists in binding nucleic acid to ZymoPURE™ columns for purification.
Catalog No. 50-125-1466 Supplier Zymo Research Corporation Supplier No. D42004150
ZymoPURE™ Binding Buffer (150 ml). Assists in binding nucleic acid to ZymoPURE™ columns for purification.