Learn More
Research Products International Corp ZymoPURE II Plasmid Gigaprep, 5 Preps
SDP
Catalog No. 501977126
Encompass
Part of the ZymoPURE Plasmid Kits collection, the ZymoPURE II Plasmid Gigaprep kit provides the fastest and simplest method available to efficiently isolate up to 10 mg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Plasmid Gigaprep kit can process up to 2.5 L of culture in less than 50 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with a vacuum instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 µg/µl) , EndoZero, vaccine-grade, and transfection-ready.
Catalog No. 50-197-7126 Supplier Research Products International Corp Supplier No. ZD4204
Part of the ZymoPURE Plasmid Kits collection, the ZymoPURE II Plasmid Gigaprep kit provides the fastest and simplest method available to efficiently isolate up to 10 mg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Plasmid Gigaprep kit can process up to 2.5 L of culture in less than 50 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with a vacuum instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 µg/µl) , EndoZero, vaccine-grade, and transfection-ready.