Learn More
Research Products International Corp ZymoPURE II Plasmid Maxiprep, 20 Preps
SDP
Catalog No. 501977183
Encompass
Part of the ZymoPURE Plasmid Kits collection, the ZymoPURE II Plasmid Maxiprep kit provides the fastest and simplest method available to efficiently isolate up to 1.2 mg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Plasmid Maxiprep kit can process up to 150 ml of culture in less than 20 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with either a vacuum or a centrifuge instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 µg/µl), EndoZero, vaccine-grade, and transfection-ready.
Catalog No. 50-197-7183 Supplier Research Products International Corp Supplier No. ZD4203
Part of the ZymoPURE Plasmid Kits collection, the ZymoPURE II Plasmid Maxiprep kit provides the fastest and simplest method available to efficiently isolate up to 1.2 mg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Plasmid Maxiprep kit can process up to 150 ml of culture in less than 20 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with either a vacuum or a centrifuge instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 µg/µl), EndoZero, vaccine-grade, and transfection-ready.