Learn More
Research Products International Corp ZymoPURE II Plasmid Midiprep, 50 Preps
SDP
Catalog No. 501977131
Encompass
Part of the ZymoPURE Plasmid Kits collection, the ZymoPURE II Plasmid Midiprep kit provides the fastest and simplest method available to efficiently isolate up to 400 µg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Midiprep kit can process up to 50 ml of culture in less than 20 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with either a vacuum or a centrifuge instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 µg/µl), EndoZero, vaccine-grade, and transfection-ready.
Catalog No. 50-197-7131 Supplier Research Products International Corp Supplier No. ZD4201
Add to Cart
Add to Cart
Part of the ZymoPURE Plasmid Kits collection, the ZymoPURE II Plasmid Midiprep kit provides the fastest and simplest method available to efficiently isolate up to 400 µg of transfection grade plasmid DNA from E. coli. Utilizing a modified alkaline lysis in conjunction with a novel binding system, the ZymoPURE II Midiprep kit can process up to 50 ml of culture in less than 20 minutes. This remarkable kit results in significantly more plasmid DNA while providing drastically reduced processing times. The plasmid DNA is bound onto a column with either a vacuum or a centrifuge instead of a lengthy gravity flow column. Additionally, there is no ethanol precipitation step required and the elution is performed using a microcentrifuge. The recovered plasmid DNA is highly concentrated (up to 3 µg/µl), EndoZero, vaccine-grade, and transfection-ready.