Learn More
Zymo Research Corporation ZymoPURE II™ Plasmid Maxiprep (20 preps)
SDP
Catalog No. NC1121573
Encompass_Preferred
ZymoPURE™ Plasmid Isolation kits provide the fastest and simplest method available to efficiently isolate transfection quality plasmid DNA from E. coli. The ZymoPURE™ Plasmid Maxiprep kit features a spin column-based method for the purification of up to 3.0 mg of high-quality plasmid DNA in less than 18 minutes. The eluted plasmid DNA is ready for immediate use, avoiding the need for subsequent precipitation steps. ZymoPURE™ technology uses a modified alkaline lysis method and features novel binding chemistry that yields highly concentrated plasmid DNA (up to 3 g/l) directly from a spin column. The wash regimen has been optimized to ensure the plasmid DNA is free of endotoxins, salt, protein, and RNA resulting in plasmid DNA suitable for sensitive applications including transfection, plasmid-mediated gene silencing, cloning, and sequencing.
Catalog No. NC1121573 Supplier Zymo Research Corporation Supplier No. D4203
ZymoPURE™ Plasmid Isolation kits provide the fastest and simplest method available to efficiently isolate transfection quality plasmid DNA from E. coli. The ZymoPURE™ Plasmid Maxiprep kit features a spin column-based method for the purification of up to 3.0 mg of high-quality plasmid DNA in less than 18 minutes. The eluted plasmid DNA is ready for immediate use, avoiding the need for subsequent precipitation steps. ZymoPURE™ technology uses a modified alkaline lysis method and features novel binding chemistry that yields highly concentrated plasmid DNA (up to 3 g/l) directly from a spin column. The wash regimen has been optimized to ensure the plasmid DNA is free of endotoxins, salt, protein, and RNA resulting in plasmid DNA suitable for sensitive applications including transfection, plasmid-mediated gene silencing, cloning, and sequencing.