Learn More
Zymo Research Corporation ZymoPURE P1 (Red), 150 ml
SDP
Catalog No. 501251448
Encompass_Preferred
ZymoPURE™ P1 (Red) (150 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.
Catalog No. 50-125-1448 Supplier Zymo Research Corporation Supplier No. D42001150
ZymoPURE™ P1 (Red) (150 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.