Learn More
Zymo Research Corporation ZymoPURE™ P1 (Red) (210 ml)
SDP
Catalog No. 501251449
Encompass_Preferred
ZymoPURE™ P1 (Red) (210 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.
Catalog No. 50-125-1449 Supplier Zymo Research Corporation Supplier No. D42001210
ZymoPURE™ P1 (Red) (210 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.