Learn More
Zymo Research Corporation ZymoPURE™ P2 (Green) (210 ml)
SDP
Catalog No. 501251455
Encompass_Preferred
ZymoPURE™ P2 (Green) (210 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.
Catalog No. 50-125-1455 Supplier Zymo Research Corporation Supplier No. D42002210
ZymoPURE™ P2 (Green) (210 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.