Learn More
Zymo Research Corporation ZymoPURE™ P3 (Yellow) (210 ml)
SDP
Catalog No. 501251461
Encompass_Preferred
ZymoPURE™ P3 (Yellow) (210 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.
Catalog No. 50-125-1461 Supplier Zymo Research Corporation Supplier No. D42003210
ZymoPURE™ P3 (Yellow) (210 ml). For use in ZymoPURE™ Plasmid Purification kits. Colored buffers permit error-free visualization and identification of complete bacterial cell lysis and neutralization.