Learn More
Zymo Research Corporation ZymoPURE Plasmid MiniPrep Kit (50 Preps)
SDP
Supplier:  Zymo Research Corporation D4209
Encompass_Preferred
The ZymoPURE™ Plasmid Miniprep Kit features a spin column-based method for the purification of up to 100 g of ultra-pure endotoxin-free plasmid DNA in less than 15 minutes. The unique spin-column design also provides zero buffer retention and a low elution volume. ZymoPURE™ technology uses a modified alkaline lysis method and features novel binding chemistry that yields highly concentrated plasmid DNA (up to 3 g/l). In addition, the wash regimen has been optimized to ensure the plasmid DNA is free of endotoxins, salt, protein, and RNA. The result is plasmid DNA suitable for transfection, restriction endonuclease digestion, bacterial transformation, PCR amplification, DNA sequencing, and other sensitive downstream applications. As an added convenience, the ZymoPURE™ Plasmid Miniprep Kit contains colored buffers that permit error-free visualization and identification of complete bacterial cell lysis and neutralization.
Catalog No. 50-125-1482