Learn More
Zymo Research Corporation ZymoPURE Wash Buffer 1 (500 ml)
SDP
Catalog No. NC2144353
Encompass_Preferred
ZymoPURE Wash 1 is designed to be used with our ZymoPURE Plasmid Purification Kits, which is the fastest and simplest method available to efficiently isolate transfection-grade plasmid DNA from E. coli.
This item is not returnable. View return policy
Catalog No. NC2144353 Supplier Zymo Research Corporation Supplier No. D42005500
ZymoPURE Wash 1 is designed to be used with our ZymoPURE Plasmid Purification Kits, which is the fastest and simplest method available to efficiently isolate transfection-grade plasmid DNA from E. coli.