Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zymosan, Saccharomyces cerevisiae

Catalog No. AAJ62876MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62876MD 250 mg
AAJ6287603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62876MD Supplier Thermo Scientific Chemicals Supplier No. J62876MD
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 58856-93-2

An insoluble preparation of yeast cell, has been shown to activate macrophages via toll-like receptor 2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

An insoluble preparation of yeast cell, has been shown to activate macrophages via toll-like receptor 2.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58856-93-2
MDL Number MFCD00082157

Specifications

Color White
Quantity 250 mg
Merck Index 14,10200
Physical Form Powder
Chemical Name or Material Zymosan, Saccharomyces cerevisiae
RTECSNumber ZI2750000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.