Learn More
Medchemexpress LLC Zymostenol-d7 | 2260669-19-8 | 99.9% | 393.70 | 1 MG
SDP
Catalog No. 5000436548
Encompass_Preferred
Zymostenol-d7 is deuterium labeled 5a-Cholest-8-en-3b-ol.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-36548 Supplier Medchemexpress LLC Supplier No. HY113345S1MG
Zymostenol-d7 is deuterium labeled 5a-Cholest-8-en-3b-ol.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.