Learn More
Avanti Polar Lipids 5α-cholesta-8,24-dien-3ß-ol, 128-33-6, MFCD00945032, 1 mg
Supplier:  Avanti Polar Lipids 700068P1MG
Encompass
Molecular Formula: C27H44O, Formula Weight: 384.638, Purity >98%.
Catalog No. NC1555040