Learn More
Zymo Research Corporation ZymoTaq™ Polymerase (200 Rxns)
SDP
Catalog No. 50444511
Encompass_Preferred
This unique product is specifically designed for the amplification of bisulfite-treated DNA for methylation detection, real-time and quantitative PCR that are SYBR Green and probe based. The product generates specific amplicons with little or no by-productformation. Simple and easy to use: heat at 95 C for 10 minutes to initiate polymerization.ZymoTaq™ DNA polymerase is a heat-activated, hot startpolymerase that has 3-terminal transferase activity. Theaddition of A overhangs to amplified DNA makes it ideal for use in TA-cloning.
Catalog No. 50-444-511 Supplier Zymo Research Corporation Supplier No. E2002
This unique product is specifically designed for the amplification of bisulfite-treated DNA for methylation detection, real-time and quantitative PCR that are SYBR Green and probe based. The product generates specific amplicons with little or no by-productformation. Simple and easy to use: heat at 95 C for 10 minutes to initiate polymerization.ZymoTaq™ DNA polymerase is a heat-activated, hot startpolymerase that has 3-terminal transferase activity. Theaddition of A overhangs to amplified DNA makes it ideal for use in TA-cloning.