Learn More
Zymo Research Corporation ZymoTaq qPCR Premix (200 Rxns)
SDP
Catalog No. 502037200
Encompass
It features a hot-start DNA polymerase and buffer system optimized for the amplification of bisulfite-treatedDNA. It also includes an intense dsDNA-specific fluorescentdye, SYTO 9, for sensitive real-time DNA quantification. This technology can also be found in the Femto Quantification Kits (Cat. No. E2005, E2006, E2007) for low copy detection of DNA from human, bacteria, or fungi.
Catalog No. 50-203-7200 Supplier Zymo Research Corporation Supplier No. E2055
It features a hot-start DNA polymerase and buffer system optimized for the amplification of bisulfite-treatedDNA. It also includes an intense dsDNA-specific fluorescentdye, SYTO 9, for sensitive real-time DNA quantification. This technology can also be found in the Femto Quantification Kits (Cat. No. E2005, E2006, E2007) for low copy detection of DNA from human, bacteria, or fungi.