Learn More
Zymo Research Corporation ZymoTaq qPCR Premix (50 Rxns)
SDP
Catalog No. 501957641
Encompass
It features a hot-start DNA polymerase and buffer system optimized for the amplification of bisulfite-treatedDNA. It also includes an intense dsDNA-specific fluorescentdye, SYTO 9, for sensitive real-time DNA quantification. This technology can also be found in the Femto Quantification Kits (Cat. No. E2005, E2006, E2007) for low copy detection of DNA from human, bacteria, or fungi.
Catalog No. 50-195-7641 Supplier Zymo Research Corporation Supplier No. E2054
It features a hot-start DNA polymerase and buffer system optimized for the amplification of bisulfite-treatedDNA. It also includes an intense dsDNA-specific fluorescentdye, SYTO 9, for sensitive real-time DNA quantification. This technology can also be found in the Femto Quantification Kits (Cat. No. E2005, E2006, E2007) for low copy detection of DNA from human, bacteria, or fungi.