Learn More
Zymo Research Corporation Zymoprep™ Yeast Plasmid Miniprep I (100 Preps) [Includes E1004 x 1: Zymolyase (1000 Units Lyophilized) w/ Storage Buffer (500µl)]
SDP
Catalog No. NC9679724
Encompass_Preferred
There is no need for glass beads, phenol, or vortexing. Instead, plasmid DNA is reliably recovered from yeast cells whether colonies, patches on plates, or liquid cultures are sampled. Plasmid yields are typically between 0.01-0.3 ng for most 2 based plasmids from 1.5 ml overnight cultures. This kit also works well with low copy number yeast plasmids. Recovered plasmid DNA is in TE buffer and can be used for E. coli transformation, Western blotting, PCR, etc.
Catalog No. NC9679724 Supplier Zymo Research Corporation Supplier No. D2001
There is no need for glass beads, phenol, or vortexing. Instead, plasmid DNA is reliably recovered from yeast cells whether colonies, patches on plates, or liquid cultures are sampled. Plasmid yields are typically between 0.01-0.3 ng for most 2 based plasmids from 1.5 ml overnight cultures. This kit also works well with low copy number yeast plasmids. Recovered plasmid DNA is in TE buffer and can be used for E. coli transformation, Western blotting, PCR, etc.