Learn More
Zymo Research Corporation Zyppy Elution Buffer, 100 ml
SDP
Catalog No. 502037052
Encompass_Preferred
Zyppy Elution Buffer is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.
Catalog No. 50-203-7052 Supplier Zymo Research Corporation Supplier No. D40365100
Zyppy Elution Buffer is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.