Learn More
Zymo Research Corporation Zyppy™ Elution Buffer (5 ml)
SDP
Catalog No. 502037055
Encompass_Preferred
Zyppy Elution Buffer is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.
Catalog No. 50-203-7055 Supplier Zymo Research Corporation Supplier No. D403655
Zyppy Elution Buffer is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.