Learn More
Zymo Research Corporation Zyppy™ Plasmid Miniprep Kit (50 Preps)
SDP
Catalog No. 50444218
Encompass_Preferred
The Zyppy Plasmid Miniprep Kit is a plasmid DNA kit that features a pellet-free modified alkaline lysis system that bypasses the normal cell-pelleting and resuspension steps common to classical plasmid preparation procedures. Simply add the uniquely formulated 7X Lysis Buffer directly to your bacterial culture, neutralize, and then purify with Zymo-Spin column technology. Additionally, the patented colored buffers included in the kit allow for easy visualization of complete cell lysis and neutralization. The Zyppy Plasmid Miniprep Kit is the fastest and easiest method available to separate plasmid DNA from E. Coli efficiently. The plasmid DNA is high quality, endotoxin-free, and well suited for use in transfection, bacterial transformation, restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications.
Catalog No. 50-444-218 Supplier Zymo Research Corporation Supplier No. D4036
The Zyppy Plasmid Miniprep Kit is a plasmid DNA kit that features a pellet-free modified alkaline lysis system that bypasses the normal cell-pelleting and resuspension steps common to classical plasmid preparation procedures. Simply add the uniquely formulated 7X Lysis Buffer directly to your bacterial culture, neutralize, and then purify with Zymo-Spin column technology. Additionally, the patented colored buffers included in the kit allow for easy visualization of complete cell lysis and neutralization. The Zyppy Plasmid Miniprep Kit is the fastest and easiest method available to separate plasmid DNA from E. Coli efficiently. The plasmid DNA is high quality, endotoxin-free, and well suited for use in transfection, bacterial transformation, restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications.