Learn More
Zymo Research Corporation Zyppy™ Wash Buffer (48 ml)
SDP
Catalog No. 50444237
Encompass_Preferred
Zyppy Wash Buffer (Concentrate) is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.
Catalog No. 50-444-237 Supplier Zymo Research Corporation Supplier No. D4036448
Zyppy Wash Buffer (Concentrate) is designed to be used with our Zyppy Plasmid Miniprep Kit, which uses a pellet-free modified alkaline lysis method to isolate ultra-pure plasmid DNA from E. coli in only 8 minutes.