Learn More
Zymo Research Corporation Zyppy-96™ Plasmid Kit (4 x 96 Preps)
SDP
Catalog No. 50444253
Encompass_Preferred
The Zyppy-96 Plasmid Miniprep allows efficient isolation of plasmid DNA from E. coli, featuring a modified alkaline lysis system that is pellet-free. This completely bypasses tedious centrifugation, pelleting, and re-suspension steps common to conventional procedures. Instead, the uniquely formulated Deep Blue Lysis Buffer is added directly to the bacterial cultures in a 96-well block. Buffer neutralization and lysate separation steps are assisted with a specially designed Neutralization/Clearing Buffer. Eluted plasmid DNA is high quality, endotoxin-free, and well-suited for use in restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications including transfection.
Catalog No. 50-444-253 Supplier Zymo Research Corporation Supplier No. D4042
The Zyppy-96 Plasmid Miniprep allows efficient isolation of plasmid DNA from E. coli, featuring a modified alkaline lysis system that is pellet-free. This completely bypasses tedious centrifugation, pelleting, and re-suspension steps common to conventional procedures. Instead, the uniquely formulated Deep Blue Lysis Buffer is added directly to the bacterial cultures in a 96-well block. Buffer neutralization and lysate separation steps are assisted with a specially designed Neutralization/Clearing Buffer. Eluted plasmid DNA is high quality, endotoxin-free, and well-suited for use in restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications including transfection.