Learn More
Zymo Research Corporation Zyppy-96™ Plasmid MagPrep Kit (4 x 96 Preps)
SDP
Catalog No. 50444286
Encompass_Preferred
The Zyppy-96 Plasmid MagBead Miniprep allows efficient isolation of plasmid DNA from E. coli, featuring a modified alkaline lysis system that bypasses tedious centrifugation, pelleting, and re-suspension steps common to conventional procedures. Instead, the uniquely formulated Deep Blue Lysis Buffer is added directly to bacterial cultures in a 96-well block. Buffer neutralization and lysate separation steps are expedited using a specially designed Neutralization Buffer. Plasmid DNA is then purified using the featured magnetic bead based technology. This straightforward procedure is compatible for either automated or manual processing. Eluted plasmid DNA is of the highest quality, endotoxin-free, and is well suited for use in restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications including transfection.
Catalog No. 50-444-286 Supplier Zymo Research Corporation Supplier No. D4101
The Zyppy-96 Plasmid MagBead Miniprep allows efficient isolation of plasmid DNA from E. coli, featuring a modified alkaline lysis system that bypasses tedious centrifugation, pelleting, and re-suspension steps common to conventional procedures. Instead, the uniquely formulated Deep Blue Lysis Buffer is added directly to bacterial cultures in a 96-well block. Buffer neutralization and lysate separation steps are expedited using a specially designed Neutralization Buffer. Plasmid DNA is then purified using the featured magnetic bead based technology. This straightforward procedure is compatible for either automated or manual processing. Eluted plasmid DNA is of the highest quality, endotoxin-free, and is well suited for use in restriction endonuclease digestion, DNA ligation, PCR, transcription, sequencing, and other sensitive downstream applications including transfection.