Learn More
Aviva Systems Biology Corp ZYX Antibody - middle region (ARP34387_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34387_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZYX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZYX. Alternate names: ESP-2, HED-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55371