Learn More
Aviva Systems Biology Corp ZYX Antibody - middle region (ARP34388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZYX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZYX. Alternate names: ESP-2, HED-2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-55372