Learn More
MSC Vermont Gage Plug Gage, 0.0140 in, 1 ea
Catalog No. NC3444086
Encompass
This Class ZZ Minus Plug and Pin Gage is a precision instrument designed for accurate measurement and calibration tasks. It is ideal for setting micrometers, performing NoGo inspections, and use with verniers and other measuring devices.
  • Precision ground to 0.0140 in diameter
  • Overall length of 2 in
  • Coating/finish of 10 micro
  • Made from steel material
  • Tolerance of 0.000200 in
  • Includes certificate of accuracy with NIST traceability
Catalog No. NC3444086 Supplier MSC Supplier No. 89001408
This Class ZZ Minus Plug and Pin Gage is a precision instrument designed for accurate measurement and calibration tasks. It is ideal for setting micrometers, performing NoGo inspections, and use with verniers and other measuring devices.
  • Precision ground to 0.0140 in diameter
  • Overall length of 2 in
  • Coating/finish of 10 micro
  • Made from steel material
  • Tolerance of 0.000200 in
  • Includes certificate of accuracy with NIST traceability