accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-39-0

IUPAC Name: (bromomethyl)benzene
Synonyms: benzyl bromide
Molecular Weight (g/mol): 171.04
Molecular Formula: C7H7Br
InChi Key: AGEZXYOZHKGVCM-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC=C1
Boiling Point 
  • (3)
  • (4)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (7)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (6)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (4)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (7)
  • (1)
  • (9)