Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Bromide (stabilized with Propylene Oxide) 98.0+%, TCI America™
SDP

Catalog No. B0411100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0411100G 100 g
B041125G 25 g
B0411500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. B0411100G Supplier TCI America Supplier No. B0411100G
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 100-39-0
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.037
MDL Number MFCD00000172
InChI Key AGEZXYOZHKGVCM-UHFFFAOYSA-N
Synonym benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
PubChem CID 7498
ChEBI CHEBI:59858
IUPAC Name bromomethylbenzene
SMILES C1=CC=C(C=C1)CBr

Specifications

Color Yellow
Boiling Point 199°C
Quantity 100 g
UN Number 1737
Formula Weight 171.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Bromide (stabilized with Propylene Oxide)
EINECSNumber (3)-2689
RTECSNumber XS7965000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.