CAS RN 100-97-0
Hexamethylenetetramine, ACS, 99+%
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 MDL Number: MFCD00006895 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Turbidity Standard, 100 NTU, Formazin, Ricca Chemical
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Hexamethylenetetramine (Crystalline/USP), Fisher Chemical
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 MDL Number: MFCD00006895 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Hexamethylenetetramine (Certified ACS), Fisher Chemical
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 MDL Number: MFCD00006895 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Hexamethylenetetramine, 99%
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Hexamethylenetetramine 99.0+%, TCI America™
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 MDL Number: MFCD00006895 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Hexamethylenetetramine, 99+%
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 MDL Number: MFCD00006895 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Turbidity, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Hexamethylenetetramine, 98.5%, contains an anticaking agent
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 MDL Number: MFCD00006895 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: methenamine,hexamethylenetetramine,hexamine,urotropine,1,3,5,7-tetraazaadamantane,hexamethylenamine,aminoform,methenamin,urotropin,formamine PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Turbidity - WP, Certified Reference Material, MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Turbidity Standard, 4000 NTU, LabChem™
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N Synonym: Formazin PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Turbidity Standard, 500 NTU, Formazin, Ricca Chemical
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Turbidity Standard, 1000 NTU, Formazin, Ricca Chemical
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
Turbidity Standard, 200 NTU, Formazin, Ricca Chemical
CAS: 100-97-0 Molecular Formula: C6H12N4 Molecular Weight (g/mol): 140.19 InChI Key: VKYKSIONXSXAKP-UHFFFAOYSA-N PubChem CID: 4101 ChEBI: CHEBI:6824 SMILES: C1N2CN3CN1CN(C2)C3
- 1
- 2