Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Turbidity Standard, 200 NTU, Formazin, Ricca Chemical
SDP

Catalog No. R8819200500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8819200500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8819200500 Supplier Ricca Chemical Company Supplier No. R8819200500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-97-0 , 10034-93-2 , 7732-18-5
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Assay Percent Range Turbidity: 200 ± 5 NTU
Physical Form Suspension
Color White
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Infinitely soluble in water
Solution Type Calibration Standard
Type Analytical Standard
Solvent or Matrix Water
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Turbity Standard, 200 NTU (Nephelometric Turbidity Units) - Formazin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.