accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONClSOONH2CH3

CAS RN 1000279-69-5

IUPAC Name: 4-[5-(4-chlorophenyl)-2-methyl-3-propanoyl-1H-pyrrol-1-yl]benzene-1-sulfonamide
Synonyms: 4-[5-(4-chlorophenyl)-2-methyl-3-propanoylpyrrol-1-yl]benzenesulfonamide
Molecular Weight (g/mol): 402.89
Molecular Formula: C20H19ClN2O3S
InChi Key: ABACVOXFUHDKNZ-UHFFFAOYSA-N
SMILES: CCC(=O)C1=C(C)N(C(=C1)C1=CC=C(Cl)C=C1)C1=CC=C(C=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)