accessibility menu, dialog, popup

UserName

Viewing profile as user:
FePPdPBrBrt-But-But-But-Bu

CAS RN 1000310-63-3

IUPAC Name: Dibromo[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
Synonyms:
Molecular Weight (g/mol): 740.66
Molecular Formula: C26H44Br2FeP2Pd
InChi Key:
SMILES: [Fe].Br[Pd++]Br.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results