accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3H3CNCH3CH3(?)

CAS RN 1000578-26-6

IUPAC Name: N,N-dimethyl-5-(1,3,5-trimethyl-1H-pyrazol-4-yl)-1,2,3,4-tetrahydronaphthalen-2-amine
Synonyms: N,N-dimethyl-5-(1,3,5-trimethylpyrazol-4-yl)-1,2,3,4-tetrahydronaphthalen-2-amine
Molecular Weight (g/mol): 283.42
Molecular Formula: C18H25N3
InChi Key: BTTOYOKCLDAHHO-UHFFFAOYNA-N
SMILES: CN(C)C1CCC2=C(C=CC=C2C1)C1=C(C)N(C)N=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)